Anti-EV71-VP4 Mouse Monoclonal Antibody [clone:M001]

Supplier: Sino Biological
40013-M001-200 40013-M001-500
103655-790EA 552.92 USD
103655-790 103655-788
Anti-EV71-VP4 Mouse Monoclonal Antibody [clone:M001]
Antibodies
This antibody was obtained from a mouse immunized with a peptide (GSQVSTQRSGSHENSNSATEK)-KLH conjugate.

Type: Primary
Antigen: Enterovirus 71 VP4 Neutralizing
Clonality: Monoclonal
Clone: M001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: EV Enterovirus 73
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR