Anti-EV71-VP1 Antibody [clone:H136]

Supplier: Sino Biological
40013-H136-500 40013-H136-200
103655-784EA 352.8 USD
103655-784 103655-786
Anti-EV71-VP1 Antibody [clone:H136]
Antibodies
This antibody was obtained from Human B cells by single-cell reverse transcription polymerase chain reaction (RT-PCR).

Type: Primary
Antigen: Enterovirus 71 VP1 Neutralizing
Clonality:
Clone: H136
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG1 (from single cell PCR)
Reactivity: EV Enterovirus 71
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR