Anti-M1 Mouse Monoclonal Antibody [clone:02]

Supplier: Sino Biological
40010-MM02-100 40010-MM02-50
103612-032EA 188.32 USD
103612-032 103655-776
Anti-M1 Mouse Monoclonal Antibody [clone:02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant H1N1 (A/Puerto Rico/8/34/Mount Sinai) M1 / Matrix Protein 1 (Catalog#40010-V07E; NP_040978.1; Ser 2-Lys 252). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Matrix Protein 1
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: H1N1 (A/Puerto Rico/8/34/Mount Sinai)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR