Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DEC-205 Mouse Monoclonal Antibody [clone: 16]

Supplier: Sino Biological

16490-MM16-50 16490-MM16-100 16490-MM16-200
103612-028 103655-764 103655-766
Anti-DEC-205 Mouse Monoclonal Antibody [clone: 16]
Anti-DEC-205 Antibody

Type: Primary
Antigen: DEC-205
Clonality: Monoclonal
Clone: Clone: 16
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty