Anti-Fgf8 Rabbit Polyclonal Antibody

Supplier: Sino Biological
16124-T32-50 16124-T32-200 16124-T32-100
103611-992EA 270.87 USD
103611-992 103588-634 103611-994
Anti-Fgf8 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human Fgf8 (rh Fgf8; Catalog#16124-HNAE; NP_149355.1; Gln23-Arg204). Fgf8 specific IgG was purified by Human Fgf8 affinity chromatography.

Type: Primary
Antigen: FGF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR