Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGFP Tag Rabbit Polyclonal Antibody

Supplier: Sino Biological

16118-RP01-400 16118-RP01-100 16118-RP01-200
103655-730 103655-728 103655-726
Anti-EGFP Tag Rabbit Polyclonal Antibody
Anti-EGFP Tag Antibody

Type: Primary
Antigen: EGFP Tag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: EGFP

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty