Anti-LILRA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
16059-RP01-200 16059-RP01-100 16059-RP01-400
103655-650EA 270.2 USD
103655-650 103655-652 103655-654
Anti-LILRA5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human LILRA5 (rh LILRA5; Catalog#16059-H08H; NP_067073.1; Met1-Arg268). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LILRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR