Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GYPA Rabbit Polyclonal Antibody

Supplier: Sino Biological

16018-RP01-400 16018-RP01-200 16018-RP01-100
103655-636EA 214.31 USD InStock
103655-636 103655-634 103655-632
Anti-GYPA Rabbit Polyclonal Antibody
Antibodies
Anti-Glycophorin A Antibody

Type: Primary
Antigen: GYPA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty