Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LILRB1 Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological

16014-MM06-200 16014-MM06-100 16014-MM06-50
103655-600EA 214.31 USD InStock
103655-600 103655-598 103611-894
Anti-LILRB1 Mouse Monoclonal Antibody [clone: 06]
Antibodies
Anti-LILRB1 Antibody

Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: Clone: 06
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty