Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KIAA1279 Rabbit Polyclonal Antibody

Supplier: Sino Biological

16011-T24-200 16011-T24-100 16011-T24-50
103611-890EA 214.31 USD InStock
103611-890 103611-888 103588-592
Anti-KIAA1279 Rabbit Polyclonal Antibody
Antibodies
Anti-KIAA1279 Antibody

Type: Primary
Antigen: KIAA1279
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty