Anti-RAET1L Rabbit Polyclonal Antibody

Supplier: Sino Biological
15759-RP01-400 15759-RP01-200 15759-RP01-100
103655-552EA 351.49 USD
103655-552 103655-554 103655-556
Anti-RAET1L Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human RAET1L (rh RAET1L; Catalog#15759-H08H; AAK91503.1; Met1-Ser217). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: RAET1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR