Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDNF Mouse Monoclonal Antibody [clone: 11]

Supplier: Sino Biological

15691-MM11-50 15691-MM11-200 15691-MM11-100
103611-840 103655-542 103655-540
Anti-CDNF Mouse Monoclonal Antibody [clone: 11]
Anti-CDNF Antibody

Type: Primary
Antigen: CDNF
Clonality: Monoclonal
Clone: Clone: 11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty