Anti-MZB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15682-T60-200 15682-T60-50 15682-T60-100
103612-428EA 270.87 USD
103612-428 103588-564 103611-830
Anti-MZB1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PERP1 / MZB1 (rh PERP1 / MZB1; Catalog#15682-H08H; Q8WU39-1; Met 1- Thr 185). PERP1 / MZB1 specific IgG was purified by Human PERP1 / MZB1 affinity chromatography.

Type: Primary
Antigen: PERP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR