Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PERP1 Rabbit Monoclonal Antibody [clone: 055]

Supplier: Sino Biological

15682-R055-100 15682-R055-50
103655-532 103612-426
Anti-PERP1 Rabbit Monoclonal Antibody [clone: 055]
Anti-Proapoptotic Caspase Adap

Type: Primary
Antigen: PERP1
Clonality: Monoclonal
Clone: Clone: 055
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty