Anti-MAPK12 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15676-T52-100 15676-T52-50 15676-T52-200
103588-560EA 142.96 USD
103588-560 103611-826 103611-824
Anti-MAPK12 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ERK3 / MAPK12 / P38-gamma (rh ERK3 / MAPK12 / P38-gamma; Catalog#15676-H20B; P53778; Met1-Leu367). ERK3 / MAPK12 / P38-gamma specific IgG was purified by Human ERK3 / MAPK12 / P38-gamma affinity chromatography.

Type: Primary
Antigen: SAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR