Anti-COX5B Rabbit Polyclonal Antibody

Supplier: Sino Biological
15660-RP01-100 15660-RP01-200 15660-RP01-400
103655-480EA 562.97 USD
103655-480 103655-476 103655-478
Anti-COX5B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human COX5B (rh COX5B; Catalog#15660-H07E; P10606; Ala32-His129). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: COX5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR