Anti-ATP6V1F Rabbit Polyclonal Antibody

Supplier: Sino Biological
15605-RP01-200 15605-RP01-400 15605-RP01-100
103655-450EA 270.2 USD
103655-450 103655-452 103655-454
Anti-ATP6V1F Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ATP6V1F (rh ATP6V1F; Catalog#15605-H09E; NP_004222.2; Met1-Arg119). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ATP6V1F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR