Anti-ATL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15483-T52-50 15483-T52-200 15483-T52-100
103611-708EA 270.87 USD
103611-708 103588-538 103611-710
Anti-ATL3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ATL3 (rh ATL3; Catalog#15483-H07E; Q6DD88; Met1-Ala445). ATL3 specific IgG was purified by Human ATL3 affinity chromatography.

Type: Primary
Antigen: ATL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR