Anti-PFDN5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15442-T16-100 15442-T16-200 15442-T16-50
103611-692EA 270.87 USD
103611-692 103611-694 103588-530
Anti-PFDN5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PFDN5 / MM-1 (rh PFDN5 / MM-1; Catalog#15442-H07E; NP_002615.2; Met1-Ala154). PFDN5 / MM-1 specific IgG was purified by Human PFDN5 / MM-1 affinity chromatography.

Type: Primary
Antigen: PFDN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR