Anti-STMN1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15440-RP01-200 15440-RP01-100 15440-RP01-400
103650-836EA 351.49 USD
103650-836 103650-834 103650-838
Anti-STMN1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human STMN1 / Stathmin 1 (rh STMN1 / Stathmin 1; Catalog#15440-H07E; NP_005554.1; Met1-Asp149). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR