Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STC1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

15411-RP01-100 15411-RP01-200 15411-RP01-400
103655-382EA 106.62 USD InStock
103655-382 103655-380 103655-384
Anti-STC1 Rabbit Polyclonal Antibody
Antibodies
Anti-STC1 Antibody

Type: Primary
Antigen: STC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty