Anti-IDI2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15292-T60-100 15292-T60-200 15292-T60-50
103611-392EA 352.03 USD
103611-392 103588-486 103611-390
Anti-IDI2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human IDI2 / IPPI2 (rh IDI2 / IPPI2; Catalog#15292-H07E; Q9BXS1; Met1-Val227). IDI2 / IPPI2 specific IgG was purified by Human IDI2 / IPPI2 affinity chromatography.

Type: Primary
Antigen: IDI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR