Anti-IDI2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15292-RP01-100 15292-RP01-200 15292-RP01-400
103655-330EA 351.49 USD
103655-330 103655-332 103655-328
Anti-IDI2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human IDI2 / IPPI2 (rh IDI2 / IPPI2; Catalog#15292-H07E; Q9BXS1; Met1-Val227). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IDI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR