Anti-RBP5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15282-T16-50 15282-T16-200 15282-T16-100
103588-482EA 142.96 USD
103588-482 103611-630 103611-632
Anti-RBP5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human RBP5 / retinol binding protein 5 (rh RBP5 / retinol binding protein 5; Catalog#15282-H07E; P82980; Met1-Arg135). RBP5 / retinol binding protein 5 specific IgG was purified by Human RBP5 / retinol binding protein 5 affinity chromatography.

Type: Primary
Antigen: RBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR