Anti-GFER Rabbit Polyclonal Antibody

Supplier: Sino Biological
15278-RP01-100 15278-RP01-400 15278-RP01-200
103655-310EA 270.2 USD
103655-310 103655-312 103655-314
Anti-GFER Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human GFER / ALR (rh GFER / ALR; Catalog#15278-H08H; NP_005253.3; Met1-Asp125). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ALR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR