Anti-TPPP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15112-T16-200 15112-T16-100 15112-T16-50
103588-460EA 142.96 USD
103588-460 103612-418 103612-420
Anti-TPPP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human TPPP2 (rh TPPP2; Catalog#15112-H07E; AAH38970.1; Met1-Lys170). TPPP2 specific IgG was purified by Human TPPP2 affinity chromatography.

Type: Primary
Antigen: TPPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR