Anti-DCUN1D1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
15059-T52-100 15059-T52-200 15059-T52-50
103611-590EA 270.87 USD
103611-590 103611-592 103588-444
Anti-DCUN1D1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human DCUN1D1 / SCCRO (rh DCUN1D1 / SCCRO; Catalog#15059-H07E; NP_065691.2; Met1-Val259). DCUN1D1 / SCCRO specific IgG was purified by Human DCUN1D1 / SCCRO affinity chromatography.

Type: Primary
Antigen: DCUN1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR