Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCLY Rabbit Polyclonal Antibody

Supplier: Sino Biological

14905-T52-50 14905-T52-100 14905-T52-200
103588-440EA 106.62 USD InStock
103588-440 103611-346 103611-348
Anti-SCLY Rabbit Polyclonal Antibody
Antibodies
Anti-SCLY Antibody

Type: Primary
Antigen: SCLY
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty