Anti-WARS Rabbit Polyclonal Antibody

Supplier: Sino Biological
14827-RP01-200 14827-RP01-100 14827-RP01-400
103655-220EA 351.49 USD
103655-220 103655-222 103655-218
Anti-WARS Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human WARS / TrpRS (rh WARS / TrpRS; Catalog#14827-H07B; P23381-1; Pro2-Gln471). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: WARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR