Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNAP-alpha Rabbit Polyclonal Antibody

Supplier: Sino Biological

14800-T54-100 14800-T54-200 14800-T54-50
103611-554EA 160.46 USD InStock
103611-554 103611-340 103588-430
Anti-SNAP-alpha Rabbit Polyclonal Antibody
Antibodies
Anti-Alpha SNAP Antibody

Type: Primary
Antigen: SNAP-alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty