Anti-ACAA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14781-RP01-100 14781-RP01-200 14781-RP01-400
103655-200EA 270.2 USD
103655-200 103655-202 103655-204
Anti-ACAA1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ACAA1 / Peroxisomal 3-oxoacyl-CoA thiolase (rh ACAA1 / Peroxisomal 3-oxoacyl-CoA thiolase; Catalog#14781-H07B; NP_001598.1; Leu27-Asn424). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ACAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR