Anti-KCNIP3 Rabbit Monoclonal Antibody [clone:038]

Supplier: Sino Biological
14683-R038-50 14683-R038-100
103655-182EA 271.4 USD
103655-182 103612-398
Anti-KCNIP3 Rabbit Monoclonal Antibody [clone:038]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CSEN / calsenilin / KCNIP3 (rh CSEN / calsenilin / KCNIP3; Catalog#14683-H07E; Q9Y2W7-1; Met1-Ile256).

Type: Primary
Antigen: CSEN
Clonality: Monoclonal
Clone: 038
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR