Anti-ETHE1 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
14681-R001-100 14681-R001-50
103655-176EA 271.4 USD
103655-176 103611-328
Anti-ETHE1 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human ETHE1 / HSCO (rh ETHE1 / HSCO; Catalog#14681-H07E; O95571; Leu13-Ala254).

Type: Primary
Antigen: ETHE1
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR