Anti-TPT1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14662-T60-200 14662-T60-100 14662-T60-50
103588-408EA 142.96 USD
103588-408 103611-316 103611-538
Anti-TPT1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human TCTP / TPT1 (rh TCTP / TPT1; Catalog#14662-H07E; Met1-Cys172). TCTP / TPT1 specific IgG was purified by Human TCTP / TPT1 affinity chromatography.

Type: Primary
Antigen: TCTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR