Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSME1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

14654-RP01-400 14654-RP01-200 14654-RP01-100
103655-162EA 214.31 USD InStock
103655-162 103655-158 103655-160
Anti-PSME1 Rabbit Polyclonal Antibody
Antibodies
Anti-PSME1 Antibody

Type: Primary
Antigen: PSME1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty