Anti-PSME2 Mouse Monoclonal Antibody [clone:06]

Supplier: Sino Biological
14640-MM06-50 14640-MM06-200 14640-MM06-100
103655-146EA 270.87 USD
103655-146 103655-148 103611-312
Anti-PSME2 Mouse Monoclonal Antibody [clone:06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSME2 / PA28b (rh PSME2 / PA28b; Catalog#14640-H07E; Met1-Tyr239). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.

Type: Primary
Antigen: PSME2
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR