Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PMVK Rabbit Polyclonal Antibody

Supplier: Sino Biological

14583-T24-50 14583-T24-100 14583-T24-200
103588-396EA 106.62 USD InStock
103588-396 103611-284 103611-286
Anti-PMVK Rabbit Polyclonal Antibody
Antibodies
Anti-PMVK Antibody

Type: Primary
Antigen: PMVK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty