Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LC3B Rabbit Polyclonal Antibody

Supplier: Sino Biological

14555-RP01-400 14555-RP01-100 14555-RP01-200
103655-036EA 214.31 USD InStock
103655-036 103655-032 103655-034
Anti-LC3B Rabbit Polyclonal Antibody
Antibodies
Anti-LC3B Antibody

Type: Primary
Antigen: LC3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty