Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EIF3K Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

14553-MM01-100 14553-MM01-200 14553-MM01-50
103655-020 103655-022 103611-184
Anti-EIF3K Mouse Monoclonal Antibody [clone: 01]
Anti-eIF3K Antibody

Type: Primary
Antigen: EIF3K
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty