Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRABP1/RBP5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

14511-T16-100 14511-T16-50 14511-T16-200
103611-150 103588-346 103611-152
Anti-CRABP1/RBP5 Rabbit Polyclonal Antibody
Anti-CRABP1 Antibody

Type: Primary
Antigen: CRABP1/RBP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty