Anti-BCCIP Rabbit Polyclonal Antibody

Supplier: Sino Biological
14442-T16-200 14442-T16-100 14442-T16-50
103588-326EA 142.96 USD
103588-326 103611-114 103611-116
Anti-BCCIP Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human BRCA2 / BCCIP (rh BRCA2 / BCCIP; Catalog#14442-HNCB; AAH09771.1; Met1-Val314). BRCA2 / BCCIP specific IgG was purified by Human BRCA2 / BCCIP affinity chromatography.

Type: Primary
Antigen: BRCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR