Anti-FHIT Rabbit Polyclonal Antibody

Supplier: Sino Biological
14434-RP01-100 14434-RP01-400 14434-RP01-200
103656-806EA 562.97 USD
103656-806 103656-804 103656-802
Anti-FHIT Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human Fragile histidine triad / FHIT (rh Fragile histidine triad / FHIT; Catalog#14434-H08E; P49789; Met1-Gln147). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: FHIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR