Anti-CKB Rabbit Polyclonal Antibody

Supplier: Sino Biological
14415-RP01-400 14415-RP01-100 14415-RP01-200
103654-942EA 351.49 USD
103654-942 103654-940 103654-944
Anti-CKB Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CKB / Creatine kinase B type (rh CKB; Catalog#14415-H07B; P12277; Pro2-Lys381). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR