Anti-GGCT Mouse Monoclonal Antibody [clone:04]

Supplier: Sino Biological
14366-MM04-200 14366-MM04-100 14366-MM04-50
103611-056EA 142.96 USD
103611-056 103654-912 103654-910
Anti-GGCT Mouse Monoclonal Antibody [clone:04]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GGCT (rh GGCT; Catalog#14366-H07E; O75223-1; Met1-Leu188). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: GGCT
Clonality: Monoclonal
Clone: 04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR