Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RRM1 Mouse Monoclonal Antibody [clone: 08]

Supplier: Sino Biological

14316-MM08-200 14316-MM08-50 14316-MM08-100
103654-870EA 214.31 USD InStock
103654-870 103611-014 103654-868
Anti-RRM1 Mouse Monoclonal Antibody [clone: 08]
Antibodies
Anti-RRM1 Antibody

Type: Primary
Antigen: RRM1
Clonality: Monoclonal
Clone: Clone: 08
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty