Anti-AARSD1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14280-T52-200 14280-T52-100 14280-T52-50
103588-284EA 142.96 USD
103588-284 103610-996 103610-994
Anti-AARSD1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human AARSD1 (rh AARSD1; Catalog#14280-H07E; Q9BTE6-2; Thr176-Glu525). AARSD1 specific IgG was purified by Human AARSD1 affinity chromatography.

Type: Primary
Antigen: AARSD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR