Anti-STX8 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14243-T26-200 14243-T26-50 14243-T26-100
103610-946EA 352.03 USD
103610-946 103610-944 103588-270
Anti-STX8 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human STX8 / Syntaxin 8 (rh STX8 / Syntaxin 8; Catalog#14243-H07H; Q9UNK0; Met1-Gly215). STX8 / Syntaxin 8 specific IgG was purified by Human STX8 / Syntaxin 8 affinity chromatography.

Type: Primary
Antigen: STX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR