Anti-PPIL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14163-T60-200 14163-T60-50 14163-T60-100
103588-256EA 142.96 USD
103588-256 103610-922 103610-920
Anti-PPIL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PPIL1 (rh PPIL1; Catalog#14163-H08E; Q9Y3C6; Met1-Gly166). PPIL1 specific IgG was purified by Human PPIL1 affinity chromatography.

Type: Primary
Antigen: PPIL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR