Anti-TSPAN7 Mouse Monoclonal Antibody [clone:01]

Supplier: Sino Biological
14031-MM01-50 14031-MM01-100 14031-MM01-200
103654-670EA 270.87 USD
103654-670 103654-672 103610-878
Anti-TSPAN7 Mouse Monoclonal Antibody [clone:01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TALLA-1 / TSPAN7 / CD231 (rh TALLA-1 / TSPAN7 / CD231; Catalog#14031-H08H; AAH18036.1; Arg113-Met213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TSPAN7
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR