Anti-PRTFDC1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14023-T48-200 14023-T48-50 14023-T48-100
103588-240EA 142.96 USD
103588-240 103611-442 103611-444
Anti-PRTFDC1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PRTFDC1 (rh PRTFDC1; Catalog#14023-H07E; Q9NRG1-1; Met1-Val225). PRTFDC1 specific IgG was purified by Human PRTFDC1 affinity chromatography.

Type: Primary
Antigen: PRTFDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR